siRNA Details
Gene Symbol: Pgpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023217.4 | NP_075706.1 | 4 | 367 | ||
GenBank mRNA | AJ278829.1 | - | 4 | 299 | ||
AK003373.1 | - | 4 | 311 | |||
AK012658.1 | - | 4 | 299 | |||
AK047835.1 | - | 4 | 311 | |||
AK049632.1 | - | 4 | 290 | |||
AK181842.1 | - | 3 | 240 | |||
BC043041.1 | - | 4 | 277 | |||
BC051938.1 | - | 4 | 296 | 5717365 | ||
BY313315.1 | - | 4 | 368 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
