siRNA Details
Gene Symbol: Cmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026844.3 | NP_081120.2 | 2 | 170 | ||
GenBank mRNA | AK002429.1 | - | 2 | 170 | ||
AK004189.1 | - | 2 | 187 | |||
AK009995.1 | - | 2 | 157 | |||
AK010073.1 | - | 2 | 157 | |||
AK013475.1 | - | 2 | 167 | |||
BC027549.1 | - | 2 | 140 | 1548292 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
