siRNA Details
Gene Symbol: Metap1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025633.3 | NP_079909.1 | 4 | 431 | ||
GenBank mRNA | AJ414378.1 | - | 1 | 294 | ||
AK005241.1 | - | 4 | 428 | |||
AK010067.1 | - | 4 | 431 | |||
AK043123.1 | - | 1 | 1217 | |||
BC051534.1 | - | 1 | 305 | 3466745 | ||
BC060739.1 | - | 3 | 1939 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
