siRNA Details
Gene Symbol: Metap1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025633.3 | NP_079909.1 | 6, 7 | 734 | ||
GenBank mRNA | AJ414378.1 | - | 3, 4 | 597 | ||
AK005241.1 | - | 6, 7 | 731 | |||
AK010067.1 | - | 6, 7 | 734 | |||
BC051534.1 | - | 3, 4 | 608 | 3466745 | ||
BC060739.1 | - | 5, 6 | 2242 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
