siRNA Details
Gene Symbol: Teddm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025634.3 | NP_079910.1 | 1 | 190 | ||
GenBank mRNA | AK009367.1 | - | 1 | 134 | ||
AK009496.1 | - | 1 | 148 | |||
AK009566.1 | - | 1 | 192 | |||
AK009632.1 | - | 1 | 152 | |||
AK009635.1 | - | 1 | 152 | |||
AK009751.1 | - | 1 | 250 | |||
AK009766.1 | - | 1 | 142 | |||
AK009777.1 | - | 1 | 134 | |||
AK009984.1 | - | 1 | 140 | |||
AK009997.1 | - | 1 | 189 | |||
AK010016.1 | - | 1 | 151 | |||
BC115655.1 | - | 1 | 141 | 40103183 | ||
BC115656.1 | - | 1 | 140 | 40103184 |
