siRNA Details
Gene Symbol: Teddm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025634.3 | NP_079910.1 | 1 | 954 | ||
GenBank mRNA | AK009367.1 | - | 1 | 898 | ||
AK009496.1 | - | 1 | 911 | |||
AK009566.1 | - | 1 | 956 | |||
AK009632.1 | - | 1 | 915 | |||
AK009635.1 | - | 1 | 916 | |||
AK009751.1 | - | 1 | 1013 | |||
AK009766.1 | - | 1 | 906 | |||
AK009777.1 | - | 1 | 898 | |||
AK009984.1 | - | 1 | 904 | |||
AK009997.1 | - | 1 | 950 | |||
AK010016.1 | - | 1 | 915 | |||
BC115655.1 | - | 1 | 905 | 40103183 | ||
BC115656.1 | - | 1 | 904 | 40103184 |
