siRNA Details
Gene Symbol: Cenpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080158.1 | NP_001073627.1 | 2 | 281 | ||
NM_025639.4 | NP_079915.1 | 3 | 282 | |||
NM_178269.3 | NP_840000.2 | 3 | 282 | |||
GenBank mRNA | AK010885.1 | - | 3 | 274 | ||
AK011191.1 | - | 3 | 259 | |||
AK011227.1 | - | 3 | 266 | |||
AK011464.1 | - | 3 | 283 | |||
AK011755.1 | - | 3 | 275 | |||
AK011793.1 | - | 3 | 268 | |||
AK017679.1 | - | 3 | 275 | |||
AK168203.1 | - | 3 | 263 | |||
AK186157.1 | - | 2 | 120 | |||
AY243536.1 | - | 3 | 243 | |||
AY243537.1 | - | 3 | 243 | |||
BC009160.1 | - | 3 | 261 | 2655787 | ||
BG276334.1 | - | 2 | 281 | |||
BY710734.1 | - | 3 | 283 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
