siRNA Details
Gene Symbol: Mis18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025642.1 | NP_079918.1 | 4 | 610 | ||
GenBank mRNA | AK011719.1 | - | 4 | 460 | ||
AK012533.1 | - | 4 | 611 | |||
AK014084.1 | - | 4 | 579 | |||
AK163310.1 | - | 4 | 644 | |||
BC079900.1 | - | 4 | 622 | 30602551 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
