siRNA Details
Gene Symbol: 1700020I14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006166.1 | - | 2 | 137 | ||
AK009929.1 | - | 2 | 142 | |||
AK011393.1 | - | 2 | 131 | |||
AK011848.1 | - | 2 | 137 | |||
AK017100.1 | - | 2 | 168 | |||
AK017322.1 | - | 2 | 131 | |||
AK029148.1 | - | 133 | ||||
AK030275.1 | - | 131 | ||||
AK147270.1 | - | 144 | ||||
AK170765.1 | - | 2 | 137 | |||
BC026913.1 | - | 2 | 132 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
