siRNA Details
Gene Symbol: 1700020I14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006166.1 | - | 2 | 149 | ||
AK009929.1 | - | 2 | 154 | |||
AK011393.1 | - | 2 | 143 | |||
AK011848.1 | - | 2 | 149 | |||
AK017100.1 | - | 2 | 180 | |||
AK017322.1 | - | 2 | 143 | |||
AK029148.1 | - | 145 | ||||
AK030275.1 | - | 143 | ||||
AK147270.1 | - | 156 | ||||
AK170765.1 | - | 2 | 149 | |||
BC026913.1 | - | 2 | 144 |

More Information
Other Species Targeted

