siRNA Details
Gene Symbol: 1700017N19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081246.1 | NP_001074715.1 | 11, 12 | 1276 | ||
GenBank mRNA | AK006074.1 | - | 10, 11 | 1246 | ||
AK014917.1 | - | 11, 12 | 1253 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
