siRNA Details
Gene Symbol: 1700017N19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081246.1 | NP_001074715.1 | 7 | 651 | ||
GenBank mRNA | AK006074.1 | - | 6 | 621 | ||
AK014917.1 | - | 7 | 628 |

More Information
Other Species Targeted

Gene Ontology Categories:
