siRNA Details
Gene Symbol: LYN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002350.3 | NP_002341.1 | 2 | 349 | ||
GenBank mRNA | AK290494.1 | - | 2 | 259 | ||
BC075001.2 | - | 1 | 92 | 30915399 | ||
BC075002.2 | - | 1 | 92 | 30915611 | ||
BC126456.1 | - | 3 | 186 | 8992172 | ||
BC126458.1 | - | 3 | 233 | 8992174 | ||
M16038.1 | - | 2 | 364 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
