siRNA Details
Gene Symbol: Tsen15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025677.3 | NP_079953.2 | 3, 4 | 450 | ||
GenBank mRNA | AK004261.1 | - | 3, 4 | 386 | ||
AK011608.1 | - | 3, 4 | 407 | |||
AK017650.1 | - | 3, 4 | 451 | |||
AK169429.1 | - | 2, 3 | 3205 | |||
BC024342.1 | - | 3, 4 | 380 | 5002659 | ||
BC145701.1 | - | 3, 4 | 417 | 40131004 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
