siRNA Details
Gene Symbol: Rpe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025683.2 | NP_079959.2 | 4 | 387 | ||
GenBank mRNA | AK007735.1 | - | 4 | 393 | ||
AK017774.1 | - | 4 | 388 | |||
AK029088.1 | - | 4 | 386 | |||
AK088152.1 | - | 4 | 388 | |||
AK150008.1 | - | 4 | 425 | |||
BC006953.1 | - | 4 | 381 | |||
BC019126.1 | - | 4 | 430 | 5008075 |
