siRNA Details
Gene Symbol: Uba5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025692.3 | NP_079968.2 | 2 | 349 | ||
GenBank mRNA | AK017787.1 | - | 2 | 349 | ||
AK077825.1 | - | 2 | 331 | |||
AK151823.1 | - | 2 | 306 | |||
AK152029.1 | - | 2 | 344 | |||
AK168907.1 | - | 2 | 343 | |||
AK206146.1 | - | 2 | 66 | |||
BC019764.1 | - | 2 | 226 | 3989119 | ||
BY768723.1 | - | 2 | 350 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
