siRNA Details
Gene Symbol: Tmem41a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025693.4 | NP_079969.1 | 4, 5 | 643 | ||
GenBank mRNA | AK007685.1 | - | 4, 5 | 638 | ||
AK013518.1 | - | 3, 4 | 468 | |||
AK160292.1 | - | 4, 5 | 564 | |||
BC011512.1 | - | 4, 5 | 623 | |||
BC019770.1 | - | 4, 5 | 598 | 5124769 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
