siRNA Details
Gene Symbol: TM4SF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014220.2 | NP_055035.1 | 3 | 594 | ||
XM_017006385.1 | XP_016861874.1 | 3 | 594 | |||
GenBank mRNA | AK093829.1 | - | 1 | 217 | ||
AK130073.1 | - | 3 | 506 | |||
AK130271.1 | - | 3 | 588 | |||
AL832780.1 | - | 1 | 689 | |||
BC008442.1 | - | 3 | 461 | 4101110 | ||
BC010166.2 | - | 3 | 410 | 4040623 | ||
BC034145.1 | - | 3 | 555 | 4618019 | ||
CR456953.1 | - | 3 | 360 | |||
M90657.1 | - | 468 |
