siRNA Details
Gene Symbol: Gapvd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025709.2 | NP_079985.2 | 16, 17 | 3394 | ||
GenBank mRNA | AK014474.1 | - | 1, 2 | 260 | ||
AK129381.1 | - | 15, 16 | 2989 | |||
AK134776.1 | - | 14, 15 | 2384 | |||
BC031478.1 | - | 13, 14 | 2439 | |||
BC032088.1 | - | 13, 14 | 2439 | |||
BC048847.1 | - | 15, 16 | 2960 | |||
BC057164.1 | - | 1, 2 | 263 | |||
BC060123.1 | - | 16, 17 | 3394 | 6408778 | ||
EF155419.1 | - | 14, 15 | 2812 |
