siRNA Details
Gene Symbol: Spryd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025716.2 | NP_079992.1 | 1, 2 | 90 | ||
GenBank mRNA | AK014623.1 | - | 1, 2 | 95 | ||
AK080726.1 | - | 1, 2 | 104 | |||
AK082745.1 | - | 1, 2 | 104 | |||
BC014757.1 | - | 1, 2 | 90 | 4923277 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
