siRNA Details
Gene Symbol: Trpd52l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025741.2 | NP_080017.1 | 1 | 1018 | ||
GenBank mRNA | AK016362.1 | - | 1 | 1007 | ||
AK016450.1 | - | 1 | 1018 | |||
BC125314.1 | - | 1 | 1018 | 40129829 | ||
BC125316.1 | - | 1 | 1018 | 40129831 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
