siRNA Details
Gene Symbol: Adat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025748.4 | NP_080024.3 | 4 | 789 | ||
GenBank mRNA | AK016938.1 | - | 4 | 804 | ||
AK021291.1 | - | 4 | 477 | |||
BC062195.1 | - | 4 | 507 | 5328466 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
