siRNA Details
Gene Symbol: Zfp474 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025749.3 | NP_080025.3 | 2 | 1281 | ||
GenBank mRNA | AK007266.1 | - | 1 | 333 | ||
AK016749.1 | - | 2 | 1282 | |||
AY350709.1 | - | 4 | 1463 | |||
BC055099.1 | - | 2 | 1277 | 6704066 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
