siRNA Details
Gene Symbol: Gid4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025757.4 | NP_080033.3 | 4 | 888 | ||
GenBank mRNA | AJ404329.1 | - | 4 | 376 | ||
AK012042.1 | - | 4 | 716 | |||
AK017120.1 | - | 888 | ||||
AK039435.1 | - | 4 | 772 | |||
AK077759.1 | - | 4 | 455 | |||
AK142753.1 | - | 4 | 690 | |||
AK164644.1 | - | 4 | 755 | |||
BC044901.1 | - | 4 | 590 | 3660276 | ||
BC046821.1 | - | 4 | 702 | 6400140 | ||
BC052418.1 | - | 4 | 730 | 6401069 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
