siRNA Details
Gene Symbol: Gskip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178613.3 | NP_848728.2 | 2 | 243 | ||
GenBank mRNA | AK017049.1 | - | 767 | |||
AK020785.1 | - | 2 | 181 | |||
AK029484.1 | - | 2 | 155 | |||
AK042664.1 | - | 2 | 204 | |||
AK079775.1 | - | 2 | 189 | |||
AK168962.1 | - | 2 | 207 | |||
AK171097.1 | - | 2 | 220 | |||
BC031494.1 | - | 2 | 211 | 3156862 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
