siRNA Details
Gene Symbol: Tspan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133681.4 | NP_598442.1 | 7 | 755 | ||
GenBank mRNA | AK007428.1 | - | 7 | 753 | ||
AK078869.1 | - | 7 | 756 | |||
AK162521.1 | - | 7 | 755 | |||
BC003448.1 | - | 7 | 695 | 2654254 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
