siRNA Details
Gene Symbol: 9030624G23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256489.1 | NP_001243418.1 | 4 | 1124 | ||
GenBank mRNA | AK018572.1 | - | 4 | 1102 | ||
AK020080.1 | - | 4 | 1128 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
