siRNA Details
Gene Symbol: Rbm22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025776.2 | NP_080052.1 | 5 | 333 | ||
GenBank mRNA | AK018452.1 | - | 5 | 356 | ||
AK045116.1 | - | 5 | 353 | |||
AK150271.1 | - | 5 | 346 | |||
AK151758.1 | - | 5 | 346 | |||
AK152252.1 | - | 5 | 346 | |||
AK165277.1 | - | 5 | 351 | |||
AK166600.1 | - | 5 | 354 | |||
AK167509.1 | - | 5 | 373 | |||
AK168919.1 | - | 5 | 334 | |||
BC042691.1 | - | 5 | 352 | |||
BC080205.1 | - | 5 | 331 | 3709760 |
