siRNA Details
Gene Symbol: Ccdc109b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293644.1 | NP_001280573.1 | 4 | 596 | ||
NM_025779.3 | NP_080055.2 | 4 | 596 | |||
GenBank mRNA | AK018492.1 | - | 4 | 473 | ||
AK150724.1 | - | 4 | 494 | |||
AK150995.1 | - | 4 | 470 | |||
AK151201.1 | - | 4 | 494 | |||
AK151915.1 | - | 4 | 507 | |||
AK152326.1 | - | 4 | 467 | |||
AK154338.1 | - | 4 | 489 | |||
BC049571.1 | - | 4 | 596 | 6588325 |
