siRNA Details
Gene Symbol: Ccdc109b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025779.3 | NP_080055.2 | 5 | 675 | ||
GenBank mRNA | AK018492.1 | - | 5 | 552 | ||
AK150724.1 | - | 5 | 573 | |||
AK150995.1 | - | 5 | 549 | |||
AK151201.1 | - | 5 | 573 | |||
AK151915.1 | - | 5 | 586 | |||
AK154338.1 | - | 5 | 568 | |||
BC049571.1 | - | 5 | 675 | 6588325 |
