siRNA Details
Gene Symbol: Thap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025780.3 | NP_080056.1 | 3 | 697 | ||
GenBank mRNA | AK018577.1 | - | 3 | 677 | ||
AK051198.1 | - | 2 | 477 | |||
AK081303.1 | - | 3 | 698 | |||
BC027502.1 | - | 3 | 605 | 1515752 |
