siRNA Details
Gene Symbol: Smim7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172396.3 | NP_765984.1 | 4 | 175 | ||
GenBank mRNA | AF140607.1 | - | 4 | 175 | ||
AK009731.1 | - | 4 | 185 | |||
AK018610.1 | - | 4 | 188 | |||
AK035786.1 | - | 4 | 176 | |||
AK148562.1 | - | 4 | 187 | |||
AK149156.1 | - | 4 | 187 | |||
AK151539.1 | - | 4 | 176 | |||
AK152451.1 | - | 4 | 176 | |||
AK152748.1 | - | 4 | 176 | |||
AK169625.1 | - | 4 | 187 | |||
BC037511.1 | - | 4 | 156 | 1512313 | ||
BC055692.1 | - | 4 | 176 | 6404961 | ||
BC060963.1 | - | 4 | 193 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
