siRNA Details
Gene Symbol: Bcs1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012324.1 | - | 6, 7 | 1092 | ||
AK078925.1 | - | 6, 7 | 1104 | |||
AK079385.1 | - | 6, 7 | 1100 | |||
AK216891.1 | - | 1, 2 | 107 | |||
BC019781.1 | - | 6, 7 | 992 | 4036047 |
