siRNA Details
Gene Symbol: Taz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173547.2 | NP_001167018.1 | 8 | 861 | ||
NM_181516.6 | NP_852657.1 | 8 | 858 | |||
GenBank mRNA | AK075615.1 | - | 8 | 815 | ||
AK076481.1 | - | 8 | 826 | |||
AK170662.1 | - | 7 | 1095 | |||
AK178858.1 | - | 5 | 276 | |||
AK183640.1 | - | 3 | 390 | |||
AK188231.1 | - | 6 | 316 | |||
AY231459.1 | - | 8 | 577 | |||
AY258041.1 | - | 8 | 577 | |||
BC015305.1 | - | 8 | 823 | 4221041 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
