siRNA Details
Gene Symbol: Wdr45b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025793.3 | NP_080069.2 | 2 | 261 | ||
GenBank mRNA | AK002352.1 | - | 2 | 222 | ||
AK013170.1 | - | 2 | 211 | |||
AK051720.1 | - | 1 | 793 | |||
AK086235.1 | - | 2 | 216 | |||
AK146101.1 | - | 2 | 262 | |||
AK151975.1 | - | 2 | 223 | |||
AK168603.1 | - | 2 | 214 | |||
AK170602.1 | - | 2 | 208 | |||
AK182151.1 | - | 60 | ||||
AK188918.1 | - | 1 | 60 | |||
AK215641.1 | - | 2 | 60 | |||
BC004595.1 | - | 2 | 205 | 3584839 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
