siRNA Details
Gene Symbol: Pmpca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173180.3 | NP_775272.1 | 3 | 327 | ||
GenBank mRNA | AK004549.1 | - | 3 | 327 | ||
AK032081.1 | - | 3 | 327 | |||
AK075965.1 | - | 3 | 328 | |||
AK077218.1 | - | 3 | 328 | |||
AK147110.1 | - | 3 | 330 | |||
AK150118.1 | - | 3 | 328 | |||
AK158990.1 | - | 3 | 295 | |||
AK161291.1 | - | 3 | 328 | |||
AK167975.1 | - | 3 | 326 | |||
AK169325.1 | - | 3 | 322 | |||
AK169342.1 | - | 3 | 322 | |||
BC010810.1 | - | 3 | 312 | 4241697 |

More Information
Other Species Targeted

Gene Ontology Categories:
