siRNA Details
Gene Symbol: Mfsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025813.3 | NP_080089.1 | 10, 11 | 966 | ||
GenBank mRNA | AK004586.1 | - | 10, 11 | 967 | ||
AK032688.1 | - | 10, 11 | 934 | |||
AK075859.1 | - | 10, 11 | 996 | |||
AK135235.1 | - | 10, 11 | 967 | |||
AK136108.1 | - | 10, 11 | 958 | |||
AK152444.1 | - | 10, 11 | 988 | |||
AK159554.1 | - | 10, 11 | 994 | |||
AK166645.1 | - | 4, 5 | 358 | |||
AK208838.1 | - | 2, 3 | 83 | |||
BC024891.1 | - | 10, 11 | 948 | 3992247 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
