siRNA Details
Gene Symbol: Riok3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024182.4 | NP_077144.2 | 6, 7 | 741 | ||
GenBank mRNA | AK004748.1 | - | 6, 7 | 833 | ||
AK131635.1 | - | 6, 7 | 745 | |||
AK134327.1 | - | 6, 7 | 833 | |||
AK147074.1 | - | 6, 7 | 742 | |||
AK167309.1 | - | 6, 7 | 837 | |||
AK170882.1 | - | 6, 7 | 821 | |||
BC002255.1 | - | 3, 4 | 271 | |||
BC033271.1 | - | 6, 7 | 779 | 4016197 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
