siRNA Details
Gene Symbol: Riok3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024182.4 | NP_077144.2 | 10 | 1251 | ||
GenBank mRNA | AK004748.1 | - | 10 | 1343 | ||
AK011110.1 | - | 3 | 335 | |||
AK131635.1 | - | 10 | 1255 | |||
AK134327.1 | - | 10 | 1343 | |||
AK147074.1 | - | 10 | 1252 | |||
AK167309.1 | - | 10 | 1347 | |||
AK170882.1 | - | 10 | 1331 | |||
BC002255.1 | - | 7 | 781 | |||
BC033271.1 | - | 10 | 1289 | 4016197 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
