siRNA Details
Gene Symbol: Acadsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025826.4 | NP_080102.1 | 8 | 1072 | ||
GenBank mRNA | AK004889.1 | - | 8 | 973 | ||
AK076535.1 | - | 8 | 973 | |||
AK131628.1 | - | 8 | 1019 | |||
AK152325.1 | - | 8 | 961 | |||
AK163322.1 | - | 4 | 553 | |||
BC011235.1 | - | 8 | 954 | |||
BC035231.1 | - | 8 | 938 | |||
BC044826.1 | - | 8 | 1005 | |||
BC054428.1 | - | 8 | 1014 | 5039627 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_013084, XM_008759865, XM_008759866, U64451, FQ210776.
Gene Ontology Categories:
