siRNA Details
Gene Symbol: Tmem107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025838.2 | NP_080114.1 | 5 | 724 | ||
NM_028336.3 | NP_082612.2 | 4 | 950 | |||
GenBank mRNA | AK002804.1 | - | 5 | 694 | ||
AK003403.1 | - | 5 | 720 | |||
AK012938.1 | - | 4 | 949 | |||
AK154279.1 | - | 5 | 722 | |||
BC096686.1 | - | 5 | 702 | 1446796 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
