siRNA Details
Gene Symbol: Sdha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023281.1 | NP_075770.1 | 3 | 316 | ||
GenBank mRNA | AF095938.1 | - | 1 | 65 | ||
AK010561.1 | - | 3 | 305 | |||
AK029520.1 | - | 3 | 316 | |||
AK034928.1 | - | 3 | 317 | |||
AK049590.1 | - | 3 | 303 | |||
AK050475.1 | - | 3 | 303 | |||
AK075990.1 | - | 3 | 300 | |||
AK145923.1 | - | 3 | 314 | |||
AK147286.1 | - | 3 | 303 | |||
AK147624.1 | - | 3 | 303 | |||
AK153085.1 | - | 3 | 323 | |||
AK162148.1 | - | 3 | 299 | |||
AK169254.1 | - | 3 | 299 | |||
AK188951.1 | - | 2 | 192 | |||
AK207923.1 | - | 2 | 192 | |||
AK209906.1 | - | 1 | 124 | |||
AK215982.1 | - | 1 | 129 | |||
AK216171.1 | - | 1 | 118 | |||
BC011301.1 | - | 3 | 276 | |||
BC031849.1 | - | 3 | 324 | 5067510 | ||
DQ402975.1 | - | 1 | 110 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
