siRNA Details
Gene Symbol: 2310030G06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025865.2 | NP_080141.1 | 3, 4 | 188 | ||
GenBank mRNA | AK007611.1 | - | 3, 4 | 204 | ||
AK007929.1 | - | 3, 4 | 177 | |||
AK009541.1 | - | 3, 4 | 195 | |||
BC027409.1 | - | 3, 4 | 188 | 4035256 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
