siRNA Details
Gene Symbol: Tmx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290751.1 | NP_001277680.1 | 6 | 724 | ||
NM_025868.4 | NP_080144.1 | 8 | 1024 | |||
GenBank mRNA | AK009759.1 | - | 7 | 712 | ||
AK075750.1 | - | 8 | 1026 | |||
AK145245.1 | - | 7 | 733 | |||
AK170587.1 | - | 7 | 721 | |||
BC013544.1 | - | 7 | 706 | 4242418 | ||
BC110994.1 | - | 7 | 707 | 5349078 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
