siRNA Details
Gene Symbol: Rassf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025886.3 | NP_080162.3 | 3 | 579 | ||
GenBank mRNA | AK002243.1 | - | 3 | 519 | ||
AK010305.1 | - | 3 | 605 | |||
BC011131.1 | - | 2 | 506 | 4206694 | ||
CK617449.1 | - | 3 | 579 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
