siRNA Details
Gene Symbol: Khdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025890.3 | NP_080166.1 | 2 | 300 | ||
GenBank mRNA | AB211060.1 | - | 2 | 237 | ||
AK010377.1 | - | 2 | 237 | |||
AK145595.1 | - | 2 | 300 | |||
AK166731.1 | - | 2 | 250 | |||
BC137808.1 | - | 2 | 208 | 8860827 | ||
BC137809.1 | - | 2 | 208 | 8860828 | ||
DQ917421.1 | - | 2 | 188 | |||
DQ917422.1 | - | 2 | 188 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
