siRNA Details
Gene Symbol: Zcchc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035509.1 | NP_001030586.1 | 4 | 896 | ||
NM_001035510.1 | NP_001030587.1 | 2 | 1367 | |||
NM_025893.2 | NP_080169.2 | 4 | 844 | |||
GenBank mRNA | AK005317.1 | - | 4 | 822 | ||
AK075607.1 | - | 1 | 448 | |||
AK134856.1 | - | 1225 | ||||
AY650116.1 | - | 1 | 326 | |||
BC017627.1 | - | 4 | 757 | 4505033 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
