siRNA Details
Gene Symbol: Cisd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025902.3 | NP_080178.1 | 3 | 425 | ||
GenBank mRNA | AK003486.1 | - | 3 | 396 | ||
AK004257.1 | - | 3 | 396 | |||
AK005184.1 | - | 3 | 384 | |||
AM162549.1 | - | 3 | 917 | |||
BC058279.1 | - | 3 | 425 | 6528467 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
