siRNA Details
Gene Symbol: Rpl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025919.2 | NP_080195.1 | 4 | 323 | ||
GenBank mRNA | AK002998.1 | - | 4 | 326 | ||
AK008422.1 | - | 4 | 325 | |||
AK011211.1 | - | 4 | 319 | |||
AK011800.1 | - | 4 | 323 | |||
AK013923.1 | - | 4 | 324 | |||
AK014593.1 | - | 2 | 1481 | |||
AK088967.1 | - | 4 | 328 | |||
AK153486.1 | - | 4 | 324 | |||
AK164445.1 | - | 4 | 324 | |||
AK169053.1 | - | 4 | 324 | |||
BC021402.1 | - | 3 | 313 | 4039711 | ||
BC025077.1 | - | 3 | 312 | 2647882 | ||
BC069896.1 | - | 3 | 306 | 6475559 |
Gene Symbol: Brf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC021918.1 | - | 1782 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
