siRNA Details
Gene Symbol: Pmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025928.3 | NP_080204.1 | 3 | 307 | ||
GenBank mRNA | AF348510.1 | - | 3 | 317 | ||
AK011174.1 | - | 3 | 309 | |||
AK012113.1 | - | 3 | 308 | |||
AK013787.1 | - | 3 | 310 | |||
AK078308.1 | - | 3 | 326 | |||
AK150603.1 | - | 3 | 324 | |||
AK168200.1 | - | 3 | 308 | |||
BC034333.1 | - | 3 | 285 | |||
BC037139.1 | - | 3 | 316 | 4952550 | ||
BC049223.1 | - | 3 | 306 | 3257320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
